Novus Biologicals
Manufacturer Code:NBP220526
Catalog # NBP220526
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the C-terminus region of humanSyntaxin-BP1. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EIEE4 FLJ37475 hUNC18 MUNC18-1 N-Sec1 NSEC1 P67 Protein unc-18 homolog 1 Protein unc-18 homolog A RBSEC1 syntaxin binding protein 1 syntaxin-binding protein 1 Unc18-1 unc-18A UNC18A UNC18neuronal SEC1; MUNC18-1; N-Sec1; neuronal SEC1; p67; Protein unc-18 homolog 1; Protein unc-18 homolog A; Syntaxin-binding protein 1; Unc-18A; Unc18-1
Gene Aliases: MUNC18-1; NSEC1; P67; RBSEC1; STXBP1; UNC18; UNC18A
UniProt ID: (Human) P61764
Entrez Gene ID: (Human) 6812
Molecular Function: membrane traffic protein membrane trafficking regulatory protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.