R&D Systems
Manufacturer Code:AF5675
Catalog # AF5675
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human Syntaxin-BP1 Ser80-Ala226 Accession # P61764 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: EIEE4 FLJ37475 hUNC18 MUNC18-1 N-Sec1 NSEC1 P67 Protein unc-18 homolog 1 Protein unc-18 homolog A RBSEC1 STXBP1 syntaxin binding protein 1 syntaxin-binding protein 1 SyntaxinBP1 UNC18 Unc18-1 unc-18A UNC18A UNC18neuronal SEC1; MUNC18-1; N-Sec1; neuronal SEC1; p67; Protein unc-18 homolog 1; Protein unc-18 homolog A; Syntaxin-binding protein 1; Unc-18A; Unc18-1
Gene Aliases: MUNC18-1; NSEC1; P67; RBSEC1; STXBP1; UNC18; UNC18A
UniProt ID: (Human) P61764
Entrez Gene ID: (Human) 6812
Molecular Function: membrane traffic protein membrane trafficking regulatory protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.