Novus Biologicals
Manufacturer Code:NBP300260H
Catalog # NB065149
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with purified recombinant Syntaxin 8 (Q9UNK0 Met1-Gly215). |
Conjugate | HRP |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CARB CIP-1-associated regulator of cyclin B syntaxin 8 syntaxin-8; CIP-1-associated regulator of cyclin B; Syntaxin-8
Gene Aliases: CARB; STX8
UniProt ID: (Human) Q9UNK0
Entrez Gene ID: (Human) 9482
Molecular Function: SNARE protein membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.