Proteintech
Manufacturer Code:122061AP150UL
Catalog # 50-172-7793
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Syntaxin 8 Fusion Protein Ag2850 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CARB STX8 syntaxin 8; CIP-1-associated regulator of cyclin B; Syntaxin-8; Syntaxin-like protein 3I35
Gene Aliases: 0610007H08Rik; 1110002H11Rik; 4930571E13Rik; CARB; STX8
UniProt ID: (Human) Q9UNK0, (Rat) Q9Z2Q7, (Mouse) O88983
Entrez Gene ID: (Human) 9482, (Rat) 59074, (Mouse) 55943
Molecular Function:
SNARE protein
membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.