Proteintech
Manufacturer Code:108411AP150UL
Catalog # 50-172-6646
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Syntaxin 6 Fusion Protein Ag1284 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: STX6 syntaxin 6; Syn6; Syntaxin-6
Gene Aliases: 2310039E05Rik; 2410005I16Rik; AA437559; AI415714; AI845861; STX6
UniProt ID: (Human) B4DR17, (Mouse) Q9JKK1, (Rat) Q63635
Entrez Gene ID: (Human) 10228, (Mouse) 58244, (Rat) 60562
Molecular Function: SNARE protein membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.