Invitrogen
Manufacturer Code:MA538156
Catalog # PIMA538156
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC2113 |
Immunogen | A synthesized peptide derived from human Syntaxin 4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: p35-2 Renal carcinoma antigen NY-REN-31 Stx4 STX4A Syn4 Syn-4 syntaxin 4 syntaxin 4A (placental) Syntaxin-4; Renal carcinoma antigen NY-REN-31; syntaxin 4A (placental); Syntaxin-4
Gene Aliases: p35-2; STX4; STX4A; Syn-4; Syn4
UniProt ID: (Human) Q12846, (Rat) Q08850, (Mouse) P70452
Entrez Gene ID: (Human) 6810, (Rat) 81803, (Mouse) 20909
Molecular Function:
SNARE protein
membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.