Proteintech
Manufacturer Code:149881AP150UL
Catalog # 50-172-9890
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Syntaxin 4 Fusion Protein Ag6973 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: p35 2 STX4 STX4A syntaxin 4; Renal carcinoma antigen NY-REN-31; syntaxin 4A (placental); Syntaxin-4
Gene Aliases: p35-2; STX4; STX4A; Syn-4; Syn4
UniProt ID: (Human) Q12846, (Rat) Q08850, (Mouse) P70452
Entrez Gene ID: (Human) 6810, (Rat) 81803, (Mouse) 20909
Molecular Function:
SNARE protein
membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.