Invitrogen
Manufacturer Code:MA527388
Catalog # PIMA527388
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI3A5 |
Immunogen | Full length human recombit protein of STX3 produced in HEK293T cell |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: STX3A syntaxin 3A syntaxin-3; syntaxin 3A; Syntaxin-3
Gene Aliases: STX3; STX3A; Syn-3
UniProt ID: (Human) Q13277, (Mouse) Q64704, (Rat) Q08849
Entrez Gene ID: (Human) 6809, (Mouse) 20908, (Rat) 81802
Molecular Function:
SNARE protein
membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.