Proteintech
Manufacturer Code:664371IG150UL
Catalog # 50-173-6863
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2D3B5 |
Immunogen | Syntaxin 1A / Syntaxin 1B Fusion Protein Ag10257 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Neuron-specific antigen HPC-1; P35A; P35B; STX1B STX1B1 STX1B2 syntaxin 1B Syntaxin 1B1 Syntaxin 1B2; Synaptotagmin-associated 35 kDa protein; syntaxin 1 a; syntaxin 1A (brain); syntaxin 1A (brain)-like; syntaxin 1B1; syntaxin 1B2; Syntaxin 2; Syntaxin-1A; Syntaxin-1B; Syntaxin-1B1; Syntaxin-1B2
Gene Aliases: GEFSP9; HPC-1; P35-1; Sap; STX1; STX1A; STX1B; STX1B1; STX1B2; Stx1bl; Stx2; SYN1A; Syn1b
UniProt ID: (Human) Q16623, (Human) P61266, (Mouse) O35526, (Rat) P32851, (Mouse) P61264, (Rat) P61265
Entrez Gene ID: (Human) 6804, (Human) 112755, (Mouse) 20907, (Rat) 116470, (Mouse) 56216, (Rat) 24923
Molecular Function:
SNARE protein
membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.