Novus Biologicals
Manufacturer Code:NBP289054
Catalog # NB036645
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | SP6 |
Immunogen | Synaptic vesicle-containing fractions immunoprecipitated from human brain homogenates using anti-human Synaptophysin mAb antibodies. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: HPC-1 Neuron-specific antigen HPC-1 p35-1 STX1P35-1 SYN1A syntaxin 1A (brain) syntaxin-1A; Neuron-specific antigen HPC-1; syntaxin 1A (brain); Syntaxin-1A
Gene Aliases: HPC-1; P35-1; STX1; STX1A; SYN1A
UniProt ID: (Human) Q16623
Entrez Gene ID: (Human) 6804
Molecular Function:
SNARE protein
membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.