Novus Biologicals
Manufacturer Code:NBP256031
Catalog # NBP256031
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:QRQLEITGRTTTSEELEDMLESGNPAIF |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HPC-1 Neuron-specific antigen HPC-1 p35-1 STX1P35-1 SYN1A syntaxin 1A (brain) syntaxin-1A; Neuron-specific antigen HPC-1; syntaxin 1A (brain); Syntaxin-1A
Gene Aliases: HPC-1; P35-1; STX1; STX1A; SYN1A
UniProt ID: (Human) Q16623
Entrez Gene ID: (Human) 6804
Molecular Function:
SNARE protein
membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.