Proteintech
Manufacturer Code:133011AP150UL
Catalog # 50-172-8636
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Syntaxin 11 Fusion Protein Ag4097 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FHL4 HLH4 HPLH4 STX11 syntaxin 11 Syntaxin 11SYX11; Syntaxin-11
Gene Aliases: 5830405C08Rik; FHL4; HLH4; HPLH4; STX11
UniProt ID: (Human) O75558, (Mouse) Q9D3G5
Entrez Gene ID: (Human) 8676, (Mouse) 74732, (Rat) 292483
Molecular Function: SNARE protein membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.