Invitrogen
Manufacturer Code:MA517617
Catalog # PIMA517617
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | SP8 |
Immunogen | Human Syntaxin |
Conjugate | FITC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Neuron-specific antigen HPC-1; P35A; Synaptotagmin-associated 35 kDa protein; syntaxin 1 a; syntaxin 1A (brain); syntaxin 1A (brain)-like; Syntaxin-1A; Syntaxin-1A Neuron-specific antigen HPC-1 STX1A STXA1 STX1
Gene Aliases: HPC-1; P35-1; Sap; STX1; STX1A; SYN1A
UniProt ID: (Human) Q16623, (Mouse) O35526, (Rat) P32851
Entrez Gene ID: (Human) 6804, (Pig) 100512594, (Mouse) 20907, (Rat) 116470
Molecular Function: SNARE protein membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.