Novus Biologicals
Manufacturer Code:NBP224630SS
Catalog # NBP224630SS
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A portion of amino acids 1-50 of human Syndecan-4 was used as the immunogen. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Amphiglycan; amphiglycan MGC22217 ryudocan Ryudocan core protein SYND4ryudocan amphiglycan syndecan 4 syndecan 4 (amphiglycan ryudocan) syndecan proteoglycan 4 syndecan-4; ryudocan amphiglycan; Ryudocan core protein; SYND4; syndecan 4 (amphiglycan, ryudocan); syndecan proteoglycan 4; Syndecan-4
Gene Aliases: SDC4; SYND4
UniProt ID: (Human) P31431
Entrez Gene ID: (Human) 6385
Molecular Function:
cytoskeletal protein
membrane-bound signaling molecule
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.