R&D Systems
Manufacturer Code:FAB29651G100UG
Catalog # FA29651G100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 305515R |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human Syndecan-2/CD362 Glu19-Gly144 Accession # AAH49836 |
Conjugate | Alexa Fluor 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Rat MonoclonalAntibody
For Research Use Only
Protein Aliases: CD362; CD362 CD362 antigen Fibroglycan heparan sulfate proteoglycan 1 cell surface-associated Heparan sulfate proteoglycan core protein HSPG HSPG1 HSPG1syndecan proteoglycan 2 SDC2 SYND2 SYND2cell surface-associated heparan sulfate proteoglycan 1 syndecan 2 syndecan-2 Syndecan2; cell surface-associated heparan sulfate proteoglycan 1; Fibroglycan; heparan sulfate proteoglycan 1, cell surface-associated; Heparan sulfate proteoglycan core protein; HSPG; SYND2; syndecan proteoglycan 2; Syndecan-2
Gene Aliases: CD362; HSPG; HSPG1; SDC2; SYND2
UniProt ID: (Human) P34741
Entrez Gene ID: (Human) 6383
Molecular Function:
cell adhesion molecule
cytoskeletal protein
defense/immunity protein
extracellular matrix glycoprotein
extracellular matrix protein
membrane-bound signaling molecule
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.