Invitrogen
Manufacturer Code:MA533237
Catalog # PIMA533237
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 197 |
Immunogen | Fusion Protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CD362; cell surface-associated heparan sulfate proteoglycan 1; Fibroglycan; heparan sulfate proteoglycan 1, cell surface-associated; heparan sulfate proteoglycan 1, cell surface-associated (fibroglycan); Heparan sulfate proteoglycan core protein; HSPG; Ryudocan/syndecan 2; SYND2; syndecan proteoglycan 2; Syndecan-2
Gene Aliases: 4833414L08Rik; AA960457; CD362; HSPG; HSPG1; SDC2; SYND2; syndecan-2
UniProt ID: (Human) P34741, (Mouse) P43407, (Rat) P34900
Entrez Gene ID: (Human) 6383, (Mouse) 15529, (Rat) 25615
Molecular Function: cell adhesion molecule cytoskeletal protein defense/immunity protein extracellular matrix glycoprotein extracellular matrix protein membrane-bound signaling molecule signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.