Novus Biologicals
Manufacturer Code:NBP267174
Catalog # NBP267174
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | JM11-21 |
Immunogen | Recombinant protein within human Syndecan 1 aa 23-300(extra). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
Protein Aliases: CD138; CD138 antigen; CD138 CD138 antigen SDC SYND1heparan sulfate proteoglycan fibroblast growth factor receptor syndecan syndecan 1 syndecan proteoglycan 1 syndecan-1; heparan sulfate proteoglycan fibroblast growth factor receptor; SYND1; syndecan proteoglycan 1; Syndecan-1
Gene Aliases: CD138; SDC; SDC1; SYND1; syndecan
UniProt ID: (Human) P18827
Entrez Gene ID: (Human) 6382
Molecular Function: cell adhesion molecule cytoskeletal protein defense/immunity protein extracellular matrix glycoprotein extracellular matrix protein membrane-bound signaling molecule signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.