Invitrogen
Manufacturer Code:MA527654
Catalog # PIMA527654
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | S275-14 |
Immunogen | Fusion protein amino acids 150-239 (Cytoplasmic C2A domain) of mouse Synaptotagmin-7 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: IPCA-7; IPCA-7 PCANAP7 Prostate cancer-associated protein 7 SYT-7 SYT7 Synaptotagmin 7 Synaptotagmin VII SytVII; Prostate cancer-associated protein 7; Protein Syt7; synaptotagmin 7; Synaptotagmin VII; Synaptotagmin-7; SytVII
Gene Aliases: AI851541; B230112P13Rik; IPCA-7; IPCA7; PCANAP7; SYT-VII; SYT7; SYTVII
UniProt ID: (Human) O43581, (Mouse) Q9R0N7, (Rat) Q62747
Entrez Gene ID: (Human) 9066, (Mouse) 54525, (Rat) 59267
Molecular Function:
membrane traffic protein
membrane trafficking regulatory protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.