Proteintech
Manufacturer Code:204721AP150UL
Catalog # 50-173-2910
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synaptotagmin-4 Fusion Protein Ag14318 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HsT1192 KIAA1342 Synaptotagmin 4 synaptotagmin IV Synaptotagmin-4 SYT4 SytIV; synaptotagmin 4; Synaptotagmin IV; Synaptotagmin-4; SytIV
Gene Aliases: HsT1192; KIAA1342; Syt3; SYT4; SytIV
UniProt ID: (Human) Q9H2B2, (Rat) P50232, (Mouse) P40749
Entrez Gene ID: (Human) 6860, (Rat) 64440, (Mouse) 20983
Molecular Function: membrane traffic protein membrane trafficking regulatory protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.