Invitrogen
Manufacturer Code:OSS00023W
Catalog # PIOSS00023W
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide from the cytoplasmic domain of rat Synaptotagmin-2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Inositol polyphosphate-binding protein; IP4-binding protein; synaptotagmin 2; Synaptotagmin II; Synaptotagmin II SytII SYT2; Synaptotagmin-2; SytII
Gene Aliases: CMS7; mKIAA4194; MYSPC; R74640; RATSYNII; SYNII; SYT2; SytII
UniProt ID: (Human) Q8N9I0, (Rat) P29101, (Mouse) P46097
Entrez Gene ID: (Human) 127833, (Rat) 24805, (Mouse) 20980
Molecular Function:
membrane traffic protein
membrane trafficking regulatory protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.