Invitrogen
Manufacturer Code:OSS00022W
Catalog # PIOSS00022W
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide from n-terminal region of rat Synaptotagmin-1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: p65; synaptotagmin 1; Synaptotagmin I; Synaptotagmin-1; Synaptotagmin-1 Synaptotagmin I SytI p65 SVP65; SytI
Gene Aliases: AW124717; G630098F17Rik; P65; Syt1; SytI
UniProt ID: (Rat) P21707, (Mouse) P46096
Entrez Gene ID: (Rat) 25716, (Mouse) 20979
Molecular Function: membrane traffic protein membrane trafficking regulatory protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.