Novus Biologicals
Manufacturer Code:NBP151925
Catalog # NBP151925
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence C-RDLQSAEKEEQEK corresponding to internal region according to NP_005630.1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: DKFZp781D2042 p65 SVP65 synaptotagmin Isynaptotagmin 1 synaptotagmin-1 SYT SytI; p65; Synaptotagmin I; Synaptotagmin-1; SytI
Gene Aliases: P65; SVP65; SYT; SYT1
UniProt ID: (Human) P21579
Entrez Gene ID: (Human) 6857
Molecular Function: membrane traffic protein membrane trafficking regulatory protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.