Proteintech
Manufacturer Code:145111AP150UL
Catalog # 50-172-9502
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synaptotagmin-1 Fusion Protein Ag5970 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: p65; P65 SVP65 Synaptotagmin 1 synaptotagmin I SYT SYT1 SytI; synaptotagmin 1; Synaptotagmin I; Synaptotagmin-1; SytI
Gene Aliases: AW124717; G630098F17Rik; P65; SVP65; SYT; SYT1; SytI
UniProt ID: (Human) P21579, (Mouse) P46096
Entrez Gene ID: (Human) 6857, (Mouse) 20979
Molecular Function:
membrane traffic protein
membrane trafficking regulatory protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.