Novus Biologicals
Manufacturer Code:NBP239100AF594
Catalog # NP23910A594
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Antibody was raised against a 17 amino acid synthetic peptide near the carboxy terminus of human SYNPO. The immunogen is located within the last 50 amino acids of SYNPO. Amino Acid Squence: SPSRAWSPRAKQAPRPS |
Conjugate | Alexa Fluor 594 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
Protein Aliases: KIAA1029 synaptopodin; Synaptopodin
Gene Aliases: KIAA1029; SYNPO
UniProt ID: (Human) Q8N3V7
Entrez Gene ID: (Human) 11346
Molecular Function: actin family cytoskeletal protein cytoskeletal protein non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.