Invitrogen
Manufacturer Code:PA541779
Catalog # PIPA541779
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human SYNJ2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: inositol phosphate 5'-phosphatase 2; inositol polyphosphate-5-phosphatase H; phosphoinositide 5-phosphatase; Synaptic inositol 1,4,5-trisphosphate 5-phosphatase 2; synaptic inositol-1,4,5-trisphosphate 5-phosphatase 2; Synaptojanin-2
Gene Aliases: AI481647; INPP5H; KIAA0348; mKIAA0348; SJ2; SYNJ2
UniProt ID: (Human) O15056, (Mouse) Q9D2G5, (Rat) O55207
Entrez Gene ID: (Dog) 484052, (Horse) 100059716, (Pig) 100516167, (Human) 8871, (Rabbit) 100355301, (Mouse) 20975, (Guinea pig) 100716353, (Rat) 84018
Molecular Function: hydrolase phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.