Novus Biologicals
Manufacturer Code:NBP220543
Catalog # NBP220543
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the N-terminus region of human Synaptojanin 2. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 3.1.3.36 inositol phosphate 5'-phosphatase 2 INPP5H KIAA0348MGC44422 Synaptic inositol-145-trisphosphate 5-phosphatase 2 synaptojanin 2 synaptojanin-2; inositol phosphate 5'-phosphatase 2; inositol polyphosphate-5-phosphatase H; phosphoinositide 5-phosphatase; Synaptic inositol 1,4,5-trisphosphate 5-phosphatase 2; synaptic inositol-1,4,5-trisphosphate 5-phosphatase 2; Synaptojanin-2
Gene Aliases: INPP5H; KIAA0348; SYNJ2
UniProt ID: (Human) O15056
Entrez Gene ID: (Human) 8871
Molecular Function:
hydrolase
phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.