Invitrogen
Manufacturer Code:MA531919
Catalog # PIMA531919
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 7H10G6 |
Immunogen | Purified recombit fragment of human SYN1 (AA: 362-511) expressed in E. coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Brain protein 4.1; synapsin 1; Synapsin I; Synapsin-1; SYNI SYN1a SYN1b; truncated synapsin 1-S
Gene Aliases: Syn-1; SYN1; Syn1-S; SYN1a; SYN1b; SYNI
UniProt ID: (Human) P17600, (Rat) P09951, (Mouse) O88935
Entrez Gene ID: (Human) 6853, (Rat) 24949, (Mouse) 20964
Molecular Function:
actin family cytoskeletal protein
cytoskeletal protein
membrane traffic protein
membrane trafficking regulatory protein
non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.