Invitrogen
Manufacturer Code:MA119332
Catalog # 01-676-088
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | SYK-01 |
Immunogen | Recombit fragment (aa 5-360) of human Syk. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C do not freeze |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: p72-Syk; p72Syk; Protein tyrosine kinase p72Syk; Spleen tyrosine kinase; Tyrosine-protein kinase SYK
Gene Aliases: p72-Syk; p72syk; ptk72; SYK; Sykb
UniProt ID: (Human) P43405, (Rat) Q64725, (Mouse) P48025
Entrez Gene ID: (Human) 6850, (Rat) 25155, (Mouse) 20963
Molecular Function:
kinase
non-receptor tyrosine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.