Invitrogen
Manufacturer Code:PA19104
Catalog # 01-674-384
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Internal sequence corresponding to amino acids C-KYLQQNRHVKDKN. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: p72-Syk; SP1 Sp1 transcription factor HGNC:11205; Spleen tyrosine kinase; Tyrosine-protein kinase SYK
Gene Aliases: p72-Syk; SYK
UniProt ID: (Human) P43405
Entrez Gene ID: (Human) 6850
Molecular Function: kinase non-receptor tyrosine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.