Invitrogen
Manufacturer Code:MA515077
Catalog # PIMA515077
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | J.33.5 |
Immunogen | Synthetic peptide corresponding to residues surrounding cysteine 60 of human Survivin |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: Apoptosis inhibitor 4; apoptosis inhibitor 4 apoptosis inhibitor survivin baculoviral IAP repeat-containing protein 5 survivin variant 3 alpha; Apoptosis inhibitor survivin; Baculoviral IAP repeat-containing protein 5; survivin; survivin variant 3 alpha; TIAP
Gene Aliases: AAC-11; AP14; API4; BIRC5; EPR-1; IAP4; survivin40; TIAP
UniProt ID: (Human) O15392, (Rat) Q9JHY7, (Mouse) O70201
Entrez Gene ID: (Human) 332, (Rat) 64041, (Mouse) 11799
Molecular Function:
enzyme modulator
protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.