Invitrogen
Manufacturer Code:PA516859
Catalog # PIPA516859
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide derived from the C-terminus of human survivin |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Apoptosis inhibitor 4; apoptosis inhibitor 4 apoptosis inhibitor survivin baculoviral IAP repeat-containing protein 5 survivin variant 3 alpha; Apoptosis inhibitor survivin; Baculoviral IAP repeat-containing protein 5; survivin; survivin variant 3 alpha; TIAP
Gene Aliases: AAC-11; API4; BIRC5; EPR-1; IAP4; survivin40; TIAP
UniProt ID: (Human) O15392, (Mouse) O70201
Entrez Gene ID: (Human) 332, (Mouse) 11799
Molecular Function: enzyme modulator protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.