Invitrogen
Manufacturer Code:PA116836
Catalog # APPA6836
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ChIP assay (ChIP) | Assay dependent |
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit human survivin full-length. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C or -80°C if preferred |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: API4 Apoptosis Inhibitor 4 Apoptosis inhibitor survivin Baculoviral IAP repeat containing 5 BIRC 5 EPR 1 IAP4 SVV TIAP; Apoptosis inhibitor 4; Apoptosis inhibitor survivin; baculoviral IAP repeat containing 5; baculoviral IAP repeat-containing 5; Baculoviral IAP repeat-containing protein 5; inhibitor of apoptosis; survivin; survivin variant 3 alpha; TIAP
Gene Aliases: AAC-11; AP14; API4; BIRC5; EPR-1; IAP4; survivin40; TIAP
UniProt ID: (Dog) Q8I009, (Cat) Q6I6F4, (Human) O15392, (Rat) Q9JHY7, (Guinea pig) E3SCZ8, (Mouse) O70201
Entrez Gene ID: (Dog) 442936, (Cat) 493835, (Human) 332, (Rat) 64041, (Guinea pig) 100527953, (Mouse) 11799, (Chinese hamster) 100689366
Molecular Function:
enzyme modulator
hydrolase
lipase
protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.