R&D Systems
Manufacturer Code:AF6471
Catalog # AF6471
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human Survivin |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: API4 apoptosis inhibitor 4 Apoptosis inhibitor survivin baculoviral IAP repeat-containing 5 Baculoviral IAP Repeat-Containing Protein 5 BIRC5 EPR-1 IAP4 survivin variant 3 alpha; Apoptosis inhibitor 4; Apoptosis inhibitor survivin; Baculoviral IAP repeat-containing protein 5; survivin variant 3 alpha
Gene Aliases: API4; BIRC5; EPR-1; IAP4
UniProt ID: (Human) O15392
Entrez Gene ID: (Human) 332
Molecular Function: enzyme modulator protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.