Novus Biologicals
Manufacturer Code:NBP274412AF488
Catalog # NB274412A48
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI6B10 |
Immunogen | Full length recombinant protein of human Suppressor of Ty 4 homolog 1 (NP_003159) produced in E.coli. |
Conjugate | Alexa Fluor 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: DRB sensitivity-inducing factor 14 kDa subunit; DRB sensitivity-inducing factor 14 kDa subunit DRB sensitivity-inducing factor small subunit DSIF p14 DSIF small subunit SPT4 SPT4HhSPT4 suppressor of Ty (S.cerevisiae) 4 homolog 1 suppressor of Ty 4 homolog 1 (S. cerevisiae) SUPT4H transcription elongation factor SPT4; DRB sensitivity-inducing factor small subunit; DSIF p14; DSIF small subunit; hSPT4; small hSpt4 subunit; suppressor of Ty 4 homolog 1; Transcription elongation factor SPT4
Gene Aliases: SPT4; SPT4H; Supt4a; SUPT4H; SUPT4H1
UniProt ID: (Human) P63272
Entrez Gene ID: (Human) 6827
Molecular Function: DNA binding protein chromatin/chromatin-binding protein nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.