Novus Biologicals
Manufacturer Code:NBP136727SS
Catalog # NBP136727SS
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2B4 |
Immunogen | Partial recombinant human Sulfatase 2 isoform b [Accession# NP_940998]. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: DKFZp313E091 EC 3.1.6 EC 3.1.6.- EC 3.1.6.14 extracellular sulfatase Sulf-2 FLJ44488 FLJ90554 hSulf-2 KIAA1247HSULF-2 MGC126411 SULF-2 sulfatase 2; Extracellular sulfatase Sulf-2; hSulf-2
Gene Aliases: HSULF-2; KIAA1247; SULF2; UNQ559/PRO1120
UniProt ID: (Human) Q8IWU5
Entrez Gene ID: (Human) 55959
Molecular Function:
hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.