R&D Systems
Manufacturer Code:MAB8380100
Catalog # MAB8380100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1018387 |
Immunogen | <i>Spodoptera frugiperda Sf 21</i> (baculovirus)-derived human Sulfamidase/SGSH protein Arg23-Leu502 Accession &num P51688 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70°C as supplied. 1 month 2 to 8°C under sterile conditions after reconstitution. 6 months -20 to -70°C under sterile conditions a |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: heparan sulfate sulfatase; heparan sulfate sulfatase HSS HSSN-sulphoglucosamine sulphohydrolase MPS3A MPS3Asulphamidase N-sulfoglucosamine sulfohydrolase SFMD SFMDEC 3.10.1.1 SGSH Sulfoglucosamine sulfamidase Sulphamidase; mucopolysaccharidosis type IIIA; N-sulphoglucosamine sulphohydrolase; Sulfoglucosamine sulfamidase; Sulphamidase
Gene Aliases: HSS; MPS3A; SFMD; SGSH
UniProt ID: (Human) P51688
Entrez Gene ID: (Human) 6448
Molecular Function: hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.