Invitrogen
Manufacturer Code:PA575165
Catalog # PIPA575165
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to mature of Substance P of human origin. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C-terminal-flanking peptide; neurokinin 1; neurokinin 2; Neurokinin A; neurokinin alpha; Neuromedin L; Neuropeptide gamma; Neuropeptide K; NKA; NPK; PPT; preprotachykinin; preprotachykinin A; protachykinin; Protachykinin-1; Protachykinin-1 PPT TAC1 SP; Substance K; Substance P; Tachykinin (substance P, neurokinin A, neuropeptide K, neuropeptide gamma); tachykinin 1; tachykinin 2; tachykinin, precursor 1 (substance K, substance P, neurokinin 1, neurokinin 2, neuromedin L, neurokinin alpha, neuropeptide K, neuropeptide gamma)
Gene Aliases: 4930528L02Rik; Hs.2563; NK-1; NK1; NK2; NKA; NKNA; NPK; PPT-A; Ppt5fl; PPTA; PPTA3; RATPPTA3; SP; TAC; TAC1; TAC2
UniProt ID: (Human) P20366, (Rat) P06767, (Mouse) P41539
Entrez Gene ID: (Human) 6863, (Rat) 24806, (Mouse) 21333
Molecular Function:
neuropeptide
peptide hormone
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.