Novus Biologicals
Manufacturer Code:NB300187
Catalog # NB300187
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Guinea pig |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide made between amino acid residues 1-11 of rat Substance P [CRPKPQQFFGLM]. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Guinea Pig Polyclonal Antibody
For Research Use Only
Protein Aliases: C-terminal-flanking peptide; neurokinin 1; neurokinin 1 neurokinin 2 neurokinin A neuromedin L neurokinin alpha neuropeptide K neuropeptide gamma) neuropeptide gamma neuropeptide K NK2 NKA NKNAneurokinin alpha NPK PPT protachykinin-1 substance K substance P TAC2neuromedin L tachykinin 2 tachykinin precursor 1 tachykinin precursor 1 (substance K substance P neurokinin 1 neurokinin 2; neurokinin 2; Neurokinin A; neurokinin alpha; Neuromedin L; Neuropeptide gamma; Neuropeptide K; NKA; NPK; PPT; preprotachykinin; protachykinin; Protachykinin-1; Substance K; Substance P; tachykinin 2; tachykinin, precursor 1 (substance K, substance P, neurokinin 1, neurokinin 2, neuromedin L, neurokinin alpha, neuropeptide K, neuropeptide gamma)
Gene Aliases: Hs.2563; NK2; NKA; NKNA; NPK; TAC1; TAC2
UniProt ID: (Human) P20366
Entrez Gene ID: (Human) 6863
Molecular Function: neuropeptide peptide hormone signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.