Abnova
Manufacturer Code:PAB9675
Catalog # 89-134-324
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic phosphopeptide corresponding to residues surrounding S515 of rat Stxbp1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic phosphopeptide of Stxbp1.
For Research Use Only
Protein Aliases: ANC18HA; Munc18-1; N-Sec1; NSEC1A; p67; Protein unc-18 homolog 1; Protein unc-18 homolog A; rbSec1; rbSec1A; rbSec1B; Sec1; Syntaxin-binding protein 1; unc-18 homolog; unc-18-1; Unc-18A; Unc18-1
Gene Aliases: ANC18HA; Munc18-1; n-sec1; nSec1; NSEC1A; rbSec1; rbSec1A; rbSec1B; Sec1; Stxbp1; Unc18-1; Unc18a
UniProt ID: (Rat) P61765
Entrez Gene ID: (Rat) 25558
Molecular Function: membrane traffic protein membrane trafficking regulatory protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.