Novus Biologicals
Manufacturer Code:NBP293410100UL
Catalog # NB028702
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 581-780 of human STRN (NP_003153.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: MGC125642 SG2NA striatin striatin calmodulin binding protein striatin calmodulin-binding protein; protein phosphatase 2 regulatory subunit B'''alpha; Striatin; striatin, calmodulin binding protein; striatin, calmodulin-binding protein
Gene Aliases: PPP2R6A; SG2NA; STRN
UniProt ID: (Human) O43815
Entrez Gene ID: (Human) 6801
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.