Invitrogen
Manufacturer Code:PA552763
Catalog # PIPA552763
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human Stomatin |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Erythrocyte band 7 integral membrane protein; Erythrocyte membrane protein band 7.2; erythrocyte membrane protein band 7.2 (stomatin); erythrocyte surface protein band 7.2; Protein 7.2b; STOM BND7 EPB7 EPB72 erythrocyte band 7 integral membrane protein erythrocyte membrane protein band 7.2 stomatin protein 7.2b; Stomatin
Gene Aliases: BND7; EPB7; EPB72; STOM
UniProt ID: (Human) P27105
Entrez Gene ID: (Human) 2040
Molecular Function: cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.