Novus Biologicals
Manufacturer Code:NBP15521420UL
Catalog # NBP15521420
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to SCP2(sterol carrier protein 2) The peptide sequence was selected from the middle region of SCP2 (NP_001007099). Peptide sequence NHKHSVNNPYSQFQDEYSLDEVMASKEVFDFLTILQCCPTSDGAAAAILA. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acetyl-CoA C-myristoyltransferase; DKFZp686C12188 DKFZp686D11188 NLTP non-specific lipid-transfer protein NSL-TP Propanoyl-CoA C-acyltransferase SCP-2 SCP-CHI SCPX SCP-X sterol carrier protein 2EC 2.3.1.176 Sterol carrier protein X; Non-specific lipid-transfer protein; NSL-TP; Propanoyl-CoA C-acyltransferase; SCOX; SCP-2; SCP-2/3-oxoacyl-CoA thiolase; SCP-2/thiolase; SCP-chi; SCP-X; SCPX; Sterol carrier protein 2; Sterol carrier protein X; Straight-chain acyl-CoA oxidase
Gene Aliases: NLTP; NSL-TP; SCP-2; SCP-CHI; SCP-X; SCP2; SCPX
UniProt ID: (Human) P22307
Entrez Gene ID: (Human) 6342
Molecular Function:
transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.