Novus Biologicals
Manufacturer Code:NBP176798SS
Catalog # NBP176798SS
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide made to an C-terminal portion of the human Stathmin 1 protein (between residues 100-149) [UniProt P16949] |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C1orf215 chromosome 1 open reading frame 215 Lag Leukemia-associated phosphoprotein p18 Metablastin MGC138869 Oncoprotein 18 Op18 OP18MGC138870 phosphoprotein 19 Phosphoprotein p19 pp17 PP19 PR22 prosolin Protein Pr22 SMNLAP18FLJ32206 stathmin stathmin 1 stathmin 1/oncoprotein 18 transmembrane protein C1orf215; Leukemia-associated phosphoprotein p18; Metablastin; Oncoprotein 18; Op18; phosphoprotein 19; Phosphoprotein p19; pp17; pp19; Prosolin; Protein Pr22; Stathmin; stathmin 1/oncoprotein 18; testicular tissue protein Li 189; transmembrane protein C1orf215
Gene Aliases: C1orf215; Lag; LAP18; OP18; PP17; PP19; PR22; SMN; STMN1
UniProt ID: (Human) P16949
Entrez Gene ID: (Human) 3925
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.