Proteintech
Manufacturer Code:111571AP150UL
Catalog # 50-172-6923
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Stathmin 1 Fusion Protein Ag1633 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C1orf215 Lag LAP18 Metablastin Oncoprotein 18 OP18 Phosphoprotein p19 PP17 PP19 PR22 Prosolin Protein Pr22 SMN Stathmin stathmin 1/oncoprotein 18 Stathmin1 STMN1; leukemia associated phosphoprotein p18; leukemia-associated cytosolic phosphoprotein stathmin; Leukemia-associated gene protein; Leukemia-associated phosphoprotein p18; Metablastin; Oncoprotein 18; Op18; phosphoprotein 19; Phosphoprotein p19; pp17; pp19; Pr22 protein; Prosolin; Protein Pr22; Stathmin; stathmin 1/oncoprotein 18; testicular tissue protein Li 189; transmembrane protein C1orf215
Gene Aliases: 19k; C1orf215; Lag; LAP18; OP18; P18; P19; Pig; PP17; Pp18; PP19; PR22; prosolin; SMN; STMN1
UniProt ID: (Human) P16949, (Rat) P13668, (Mouse) P54227
Entrez Gene ID: (Human) 3925, (Rat) 29332, (Mouse) 16765
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.