Abnova
Manufacturer Code:MAB2097
Catalog # 89-110-818
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 5F6.F1 |
Immunogen | Synthetic phosphopeptide corresponding to residues surrounding Y694 of mouse Stat5a. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against synthetic phosphopeptide of Stat5a.
For Research Use Only
Protein Aliases: AA959963; Mammary gland factor; mammary gland factor STAT5A; Signal transducer and activator of transcription 5A; STAT5
Gene Aliases: AA959963; Mgf; Mpf; STAT5; Stat5a
UniProt ID: (Mouse) P42230
Entrez Gene ID: (Mouse) 20850
Molecular Function:
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.