Novus Biologicals
Manufacturer Code:NBP297046AF405
Catalog # NB080813
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with purified recombinant Stanniocalcin 1/STC-1 (NP_003146.1 Met1-Ala247). |
Conjugate | Alexa Fluor 405 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: stanniocalcin 1 stanniocalcin-1 STCSTC-1; Stanniocalcin-1; STC-1
Gene Aliases: STC; STC1
UniProt ID: (Human) P52823
Entrez Gene ID: (Human) 6781
Molecular Function: peptide hormone signaling molecule transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.