R&D Systems
Manufacturer Code:AF3645SP
Catalog # AF3645SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human Stabilin-2 Gly2198-Val2295 Accession # Q8WWQ8 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: 190 kDa form stabilin-2; 190 kDa hyaluronan receptor for endocytosis; CD44-like precursor FELL; CD44-like precursor FELL DKFZP434E0321 FAS1 EGF-like and X-link domain containing adhesion molecule-2 FAS1 EGF-like and X-link domain-containing adhesion molecule 2 fasciclin egf-like laminin-type egf-like and link domain-containing scavengerreceptor-2 Fasciclin EGF-like laminin-type EGF-like and link domain-containing scavengerreceptor 2 FEEL2 FEEL-2 FELE-2 FELL FELL-2 FEX2 HARE HAREFELL2 hepatic hyaluronan clearance receptor Hyaluronan receptor for endocytosis hyaluronic acid receptor for endocytosis STAB2 STAB-2 stabilin 2 stabilin-2 Stabilin2; FAS1 EGF-like and X-link domain containing adhesion molecule-2; FAS1 EGF-like and X-link domain-containing adhesion molecule 2; fasciclin egf-like, laminin-type egf-like, and link domain-containing scavenger receptor-2; Fasciclin, EGF-like, laminin-type EGF-like and link domain-containing scavenger receptor 2; FEEL-2; hepatic hyaluronan clearance receptor; Hyaluronan receptor for endocytosis; hyaluronic acid receptor for endocytosis; STAB2; Stabilin-2
Gene Aliases: FEEL2; FELE-2; FELL; FELL2; FEX2; HARE; SCARH1; STAB2
UniProt ID: (Human) Q8WWQ8
Entrez Gene ID: (Human) 55576
Molecular Function: extracellular matrix protein extracellular matrix structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.