Test
Novus Biologicals
Manufacturer Code:NBP275742V
Catalog # NB008769
Quantity:
Shared lists are a new way to save products. Create a list for each of your projects to share with collaborators like your lab manager. Then select and buy products as a team.
![]()
Creating a shared list
Click "Save to list" to create your first list, give it a name, and add the product. Access your shared lists in the Account link on Thermofisher.com. |
![]()
Collaborate
You can invite collaborators like your lab manager or procurement staff to any shared list. They can add more items, leave comments and complete the final purchase. |
![]()
Export as a spreadsheet
Export your list as a spreadsheet for easy documentation or hand off to your lab’s designated buyer. |
Shared lists are a new way to save products. Create a list for each of your projects to share with collaborators like your lab manager. Then select and buy products as a team.
Click "Save to list" to create your first list, give it a name, and add the product. Access your shared lists in the Account link on Thermofisher.com.
You can invite collaborators like your lab manager or procurement staff to any shared list. They can add more items, leave comments and complete the final purchase.
Export your list as a spreadsheet for easy documentation or hand off to your lab’s designated buyer.
Shared lists are a new way to save products. Create a list for each of your projects to share with collaborators like your lab manager. Then select and buy products as a team.
![]()
Creating a shared list
Click "Save to list" to create your first list, give it a name, and add the product. Access your shared lists in the Account link on Thermofisher.com. |
![]()
Collaborate
You can invite collaborators like your lab manager or procurement staff to any shared list. They can add more items, leave comments and complete the final purchase. |
Export your list as a spreadsheet for easy documentation or hand off to your lab’s designated buyer.
Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | STAR/2154 |
Immunogen | Recombinant fragment (around aa 39-108) of human STAR protein (exact sequence is proprietary) |
Conjugate | DyLight 405 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: cholesterol trafficker mitochondrial steroid acute regulatory protein StAR StARD1 STARD1StAR-related lipid transfer (START) domain containing 1 START domain containing 1 START domain-containing protein 1 steroid acute regulatory protein steroidogenic acute regulator steroidogenic acute regulatory protein steroidogenic acute regulatory protein mitochondrial
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.