Abnova
Manufacturer Code:PAB15750
Catalog # 89-117-815
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant GST fusion protein corresponding to 292 amino acids (Q2097-K2388) of rat Sptbn2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C on dry atmosphere. After reconstitution with 200&muL distilled water,store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against partial recombinant Sptbn2.
For Research Use Only
Protein Aliases: beta SpIII sigma 1; beta-III spectrin; beta-spectrin 3; glutamate transporter EAAT4-associated protein 41; spectrin beta 3; spectrin beta chain, brain 2; spectrin beta chain, non-erythrocytic 2; spectrin, non-erythroid beta chain 2; spectrin-like protein GTRAP41; SPNB-3; Spnb3
Gene Aliases: Spnb3
Entrez Gene ID: (Rat) 29211
Molecular Function: actin family cytoskeletal protein cytoskeletal protein non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.