Novus Biologicals
Manufacturer Code:NBP29372220UL
Catalog # NB029308
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1500-1726 of human SUPT6H (NP_003161.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: emb-5 hSPT6 KIAA0162MGC87943 SPT6HSPT6 suppressor of Ty (S.cerevisiae) 6 homolog suppressor of Ty 6 homolog (S. cerevisiae) Tat-cotransactivator 2 protein Tat-CT2 protein transcription elongation factor SPT6; Histone chaperone suppressor of Ty6; hSPT6; suppressor of Ty 6 homolog; Tat-cotransactivator 2 protein; Tat-CT2 protein; Transcription elongation factor SPT6
Gene Aliases: emb-5; KIAA0162; SPT6; SPT6H; SUPT6H
UniProt ID: (Human) Q7KZ85
Entrez Gene ID: (Human) 6830
Molecular Function:
DNA binding protein
chromatin/chromatin-binding protein
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.